Would you like to log in again?
COLUMN CHIRAL HYPERSIL L51 10U 250X4.6MM
Supplier: Thermo Scientific
Hypersil* HPLC Column 4.6x250mm^ Preparative column and Analytical column, Standard, Chiral, Max. Pressure: 3700 psi (250 bar), Particle Size: 10 um, Pore Size: 1000A, Surface Area: 30m2/g, USP Type: L51, Stationary Phase: Amylose-tris-(3, 5-dimethylphenyl-carbamate), Dimension: 4.6x250mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN CHIRAL HYPERSIL L51 10U 250X4.6MM 76331-914 42110-254630 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty