Would you like to log in again?
H2O2 STERILIZATION PREP FACTORY INSTALLD
Supplier: CARON PRODUCTS & SERVICE
H2O2 sterilization prep, factory installed^ H2O2 sterilization prep, factory installed. For use in 7404-10 Series incubators. Includes H2O2 sterilization module connections, upgraded system wiring, and control software & hardware. STER301 H2O2 sterilization module sold separately
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
H2O2 STERILIZATION PREP FACTORY INSTALLD 76332-016 STER307 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty