Would you like to log in again?
Guard Column Holders for Preparative HPLC Columns
Supplier: Thermo Scientific
887-031 887-020 891-030 887-030 891-031 885-010
76332-128EA 454.72 USD InStock
76332-128 76332-136 76332-126 76332-134 76332-130 76332-084
Guard Column Holders for Preparative HPLC Columns
Guard Column Holders
Using preparative guards is a proven and cost effective way to extend the lifetime of your preparative columns. Thermo Scientific™ Preparative Guard Column Holders allow for easy assembly and connection of the guard column to the preparative column.
A range of guard holder sizes are offered, which are all in a convenient stand-alone format that requires connections from the LC inlet and outlet to the preparative LC column.
Preparative guard holders are available in ¹/₁₆ or ¹/₈" connection fittings in the larger guard sizes to be compatible with your Preparative HPLC system hardware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty