Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hypersil GOLD™ PREP aQ HPLC Columns

Supplier: Thermo Scientific
Hypersil GOLD™
25305-029323A 25305-159270XL 25305-109370A 25305-159370A 25305-159270A 25305-259070A 25305-259570A 25305-109070A 25305-029323B 25305-019323B 25305-109570XL 25305-109270XL 25305-259370XL 25305-259570XL 25305-259370A 25305-109270A 25305-159070A 25305-259270A 25305-159570XL 25305-259270XL 25305-109370XL 25305-109570A 25305-159570A 25305-019023A 25305-018823A 25305-159370XL 25305-019323A
76332-228EA 19462.49 USD InStock
76332-228 76332-226 76332-246 76332-224 76332-222 76332-244 76332-388 76332-220 76332-386 76332-240 76332-384 76332-382 76332-380 76332-218 76332-216 76332-238 76332-378 76332-236 76332-214 76332-376 76332-234 76332-374 76332-232 76332-394 76332-230 76332-392 76332-390
Hypersil GOLD™ PREP aQ HPLC Columns
Chromatography Columns
Thermo Scientific Hypersil GOLD™ aQ Polar endcapped C18 preparative columns are perfect for challenging reversed phase separations using highly aqueous mobile phases. The columns come in a wide range of dimensions and an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.

  • Stability in highly aqueous mobile phases
  • Alternative selectivity with polar endcapped C18 stationary phase
  • Standard threaded and standard flange hardware

Alternative selectivity with polar endcapped C18 stationary phase:
Polar functional group in endcap provides further interaction mechanism for separation of polar compounds
Yields superior retention of polar compounds
C18 (USP L1) selectivity

Stability in highly aqueous mobile phases:
Applicable in 100% aqueous mobile phases
No loss of performance or stability

Standard threaded and standard flange hardware:
Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use
Standard threaded hardware for column int.Ø. <40 mm
Standard flange hardware for column int.Ø. ≥40 mm
Standard hardware can be modified on request to be compatible with SFC

XtendedLife hardware:
Improves column lifetime by incorporating an internal Dynamic axial compression (DAC) system, ensuring the media bed is continuously packed tightly
Compatible with SFC and is available in dimensions up to 50 mm internal diameter

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty