Would you like to log in again?
COLUMN PREP HYPERCARB STD 7UM 100X30MM
Supplier: Thermo Scientific
Hypercarb* HPLC Column 30x100mm^ Preparative column, Standard, Reversed Phase, Carbon Load: 100%, Max. Pressure: 2200 psi (150 bar), Particle Size: 7 um, pH: 0 to 14, Pore Size: 250A, Surface Area: 120m2/g, USP Type: L109, Stationary Phase: Porous Graphitic Carbon, Dimension: 30x100mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN PREP HYPERCARB STD 7UM 100X30MM 76332-274 35007-109370A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty