Would you like to log in again?
COLUMN PREP SYNCRONIS C18 5UM 150X20MM
Supplier: Thermo Scientific
Syncronis* HPLC Column 20x150mm^ Preparative column, Standard, Reversed Phase, Carbon Load: 16%, Max. Pressure: 4300 psi (300 bar), Particle Size: 5 um, pH: 2 to 9, Pore Size: 100A, Surface Area: 320m2/g, Temperature: 60deg C, USP Type: L1, Stationary Phase: C18, Dimension: 20x150mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN PREP SYNCRONIS C18 5UM 150X20MM 76332-288 97105-159270A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty