Would you like to log in again?
BetaSil™ C18 HPLC Preparative Columns
Supplier: Thermo Scientific
BetaSil™
70105-259070A 70105-159270A
76332-364EA 2688.66 USD InStock
76332-364 76332-362
BetaSil™ C18 HPLC Preparative Columns
Chromatography Columns
Deliver the high surface area with high bonded phase coverage for scaling up purification needs with Thermo Scientific™ BetaSil™ C18 preparative HPLC columns.
Standard threaded and standard flange hardware:
Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use
Standard threaded hardware for column int.Ø <40 mm
Standard flange hardware for column int.Ø ≥40 mm
Standard hardware can be modified on request to be compatible with SFC
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty