Would you like to log in again?
BioBasic™ 18 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
72105-254630 72105-052130 72105-102130 72105-014001 72105-011001 72105-252130 72105-151030 72105-154630 72105-259270A 72105-152130 72105-101030 72105-051030 72105-012101
10043-504EA 1117.55 USD InStock
10043-504 10043-478 10043-456 10043-446 10043-502 10043-448 10043-476 10046-484 10046-472 10046-490 76332-366 10046-466 10046-464
BioBasic™ 18 HPLC Columns, Thermo Scientific
Chromatography Columns
For outstanding separation of small-to-medium size peptides, choose Thermo Scientific™ BioBasic™ 18 HPLC Columns.
BioBasic reversed-phase columns provide superior chromatography for life science applications because the extra dense bonding chemistry used for these packings produces a highly stable, reproducible surface for reliable results.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm ID require the 1mm UNIGUARD Holder (10046-746), 2.1/3 mm ID require the 2/3 mm UNIGUARD Holder (10046-748) and 4/4.6 mm ID require the 4/4.6 mm UNIGUARD Holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty