Would you like to log in again?
Hypersil GOLD™ C4 HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
Hypersil GOLD™
25503-154630 25505-259270XL 25503-013001 25505-254630 25503-052130 25505-109270XL 25502-102130 25505-259070A 25502-052130 25502-152130 25503-012101 25505-154630
10041-560EA 1263.96 USD InStock
10041-560 10041-562 10041-596 10041-554 76332-398 10041-578 76332-396 10041-568 10045-660 76420-876 10041-592 10041-570
Hypersil GOLD™ C4 HPLC Columns, Thermo Scientific
Chromatography Columns
For very hydrophobic analytes, achieve faster separations with excellent peak shapes using Thermo Scientific™ Hypersil GOLD™ C4 HPLC columns.
Featuring a lower hydrophobicity than C18 or C8 columns, Hypersil GOLD C4 columns provide outstanding sensitivity and high efficiency.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty