Would you like to log in again?
Syncronis™ PREP HILIC HPLC Columns
Supplier: Thermo Scientific
Syncronis™
97505-259270XL 97505-019023A 97505-259570A 97505-259270A 97505-029323B 97505-259570XL 97505-259370A 97505-259370XL 97505-029323A 97505-019323A 97505-019323B
76332-426EA 24362.27 USD InStock
76332-426 76332-434 76332-432 76332-430 76332-428
Syncronis™ PREP HILIC HPLC Columns
Chromatography Columns
Syncronis™ HILIC preparative HPLC columns for polar and hydrophilic analytes.
  • Zwitterionic bonded phase enhances retention of polar and hydrophilic analytes
  • Enhanced sensitivity with MS detection
  • Highly organic mobile phase facilitates desolvation in ESI-MS
Highly pure, high surface area silica and dense stationary phase coverage minimize secondary interactions and deliver reproducible high-resolution separations. The zwitterionic stationary phase results in a neutral, highly polar surface and provides optional selectivity to C18 columns without ion-pairing or derivatization. These columns come in a wide range of dimensions and an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed.
Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use. Standard threaded hardware for column I.D. <40 mm. Standard flange hardware for column I.D. ≥40 mm. Standard hardware can be modified on request to be compatible with SFC.
Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly. Compatible with SFC and is available in dimensions up to 50 mm I.D.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty