Would you like to log in again?
Whatman™ Cyclopore PC Membranes, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Cyclopore™
7060-4704 10417606 10417712 7060-2501 7060-2511 10419412 10418450 7060-2504 7060-2513 7060-4701 7060-2502 7060-4710 7060-4710 10417612 7060-4714 7060-4701 7060-2514 10417706 10419406 10418552 7060-4713 7060-2501 10419406 10419412 10418550 7060-2502 7060-2514 7060-4704 10417701 7060-4713 7060-2511 7060-4714 7060-4712 7060-2513 7060-4712
28350-084PK 236.14 USD InStock
28350-084 CA28350-096 CA76881-914 10147-974 10035-748 CA76881-912 CA28350-090 76332-646 CA11014-000 76881-912 10147-924 CA10147-976 76457-212 76457-214 CA10147-974 10035-752 CA89187-136 CA28350-084 28350-090 10128-890 76457-216 CA10128-888 76881-914 CA11013-182 76332-648 28350-096 76332-654 76332-652 76332-650 10035-746 10147-976 CA11013-618 10128-888 CA10128-890 CA10147-924
Whatman™ Cyclopore PC Membranes, Whatman products (Cytiva)
Filters Membrane Filters
Cytiva's Whatman Cyclopore polycarbonate membranes are true pore size microporous membranes featuring the sharp cut-off and reproducible microfiltration characteristics of track-etched membranes.
  • Low affinity for stains improves visibility under a microscope
  • Low nonspecific protein binding minimizes sample loss of proteinaceous samples
  • Narrow micropore size distribution and high pore density support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • No particle shedding enables clean filtrate
  • Biologically inert
Whatman Cyclopore track-etched membrane filter circles are suitable for a wide variety of applications, from general filtration, including cross-flow filtration (tangential flow filtration), to air and water monitoring, microorganism analysis, and many healthcare applications, including immunodiagnostics.
Cytiva business offers a variety of Whatman track-etched membranes. Cyclopore comes in clear and black colors. Nuclepore track-etched membrane options are available for organic halide adsorption, electron microscopy, and other specific applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty