Would you like to log in again?
Steri-Wrap® III Non-Woven Sterilization Wrap, Propper Manufacturing
Supplier: Propper
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
31303600 31304810 31302440 31301800
76334-496CS 701.03 USD InStock
76334-496 76334-494 76333-060 76334-492
Steri-Wrap® III Non-Woven Sterilization Wrap, Propper Manufacturing
Sterilization Wraps
Steri-Wrap® III is a heavy weight wrap suitable for your heaviest instruments in steam and ethylene oxide sterilization cycles.
  • Cellulose based wrap for steam and EO sterilization
  • Efficient microbial barrier, high-level of protection against fluids and particles
  • Highly permeable meaning no wet packs
  • Variety of strength of fabric to meet your needs
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty