Would you like to log in again?
Empore™ Solid Phase Extraction Cartridges, CDS Analytical
Supplier: CDS Analytical
Empore®
98-0604-0198-5 98-0604-0199-3 98-0604-0188-6 98-0604-0191-0 98-0604-0203-3 98-0604-0197-7
76333-126CS 591.72 USD InStock
76333-126 76333-116 76333-118 76333-122 76333-124 76333-120
Empore™ Solid Phase Extraction Cartridges, CDS Analytical
Chromatography Columns
Improve quality of data and save time and money with Empore SPE cartridges.
  • Eliminate evaporation steps with the lowest sample and elution volumes
  • Avoid clogging of frits, commonly caused by loose-packed SPE cartridges, to reduce LC-MS downtime
  • Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
  • High reproducibility – Uniform particle bed improves lot-to-lot consistency
  • Universal compatibility – Cartridge design features a standard luer tip fitting compatible with standard cartridge manifolds
The unique matrix of the Empore membrane allows for optimized elution volume, which improves sensitivity, reproducibility, provides cleaner chromatograms, and eliminates evaporation time to improve throughput. Additionally, the Empore membrane is resistant to clogging and will not contribute to degradation of instrument column lifetime.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty