Would you like to log in again?
DuoClick™ Specimen Containers with Temperature Strip, Parter Medical Products
Supplier: Parter Medical Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
143024 243024
76333-278CS 217.14 USD InStock
76333-278 76333-280
DuoClick™ Specimen Containers with Temperature Strip, Parter Medical Products
Sample Containers General Purpose Containers
The DuoClick™ system helps ensure an audible notification of correct closure. Specialty containers with temperature strip allows for temperature monitoring of specimen in drug screening and toxicology applications.
  • DuoClick™ lid alerts user of correct, leak-proof closure
  • Leak Resistant
  • Graduation markings for accurate volume measurement
Specimen containers are the ideal choice for collection and transport of specimens in microbiology, urinalysis, pathology/histology, and drug testing laboratories. All containers are available sterile or non-sterile, and with many lids, labeling, and packaging options. Simply turn the lid until you hear the two 'clicks' to fully ensure 95 kPa compliance.
Certifications: 95 kPa certified.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty