Would you like to log in again?
VWR® Microtube Racks, 64-Place
Supplier: VWR
VWR®
5310-00-22AV1 5310-29-22AV1
76333-294CS 495.47 USD InStock
76333-294 76333-282
VWR® Microtube Racks, 64-Place
Racks Tube Racks
The 64-place freezer storage rack is designed to replace 51/4×51/4" cardboard freezer storage boxes.
  • Rack holds up to 64 tubes in an 8×8 matrix
  • Produced from copolymer polypropylene
  • Withstands limited autoclaving (up to 15 minutes) as well as deep freezer storage (down to –90 °C)
  • Both sides are labeled with a large easy-to-read 1 to 8 by A-H matrix
Designed to be reversible; one side has holes suitable for the larger tubes the other has holes suitable for smaller size microtubes. The hole spacing is greater than with the standard 80-place rack making it possible to store tubes in all 64 holes simultaneously.
Great substitute for cardboard or metal freezer racks:
Side A: Holds 64×1.5/2.0 ml tubes
Side B: Holds 64×0.5 ml tubes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty