Would you like to log in again?
VWR® Microtube Racks, 96-Place
Supplier: VWR
VWR®
5322-00-21AV1 5322-34-21AV1 5322-35-21AV1 5322-33-21AV1 5322-36-21AV1 5322-32-21AV1 5322-39-21AV1 5322-31-21AV1
76333-314CS 183.91 USD InStock
76333-314 76333-304 76337-380 76333-310 76333-312 76333-306 76337-382 76333-308
VWR® Microtube Racks, 96-Place
Racks Tube Racks
Double-sided racks hold 1.7 and 2.0 ml tubes on one side and 0.5 ml tubes on the reverse side.
  • Rack for use with 0.5, 1.7, and 2.0 ml microcentrifuge tubes and 2.0 ml cryogenic storage vials
  • Rack and clear cover (sold separately) are autoclavable and freezable
Racks are available in eight colors and an assorted color pack with one of each color: blue, green, orange, red, and yellow.
Ordering information: Clear cover sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty