Would you like to log in again?
4-Port Vacuum Manifold, Left Handed Trap Assembly, Double Bank, Airfree, Schlenk, Chemglass
Supplier: Chemglass
AF-0451-02
76333-328EA 6255.39 USD InStock
76333-328
4-Port Vacuum Manifold, Left Handed Trap Assembly, Double Bank, Airfree, Schlenk, Chemglass
Manifolds
Similar to the AF-0450 5-port line, except that the line is constructed using 0 to 4 mm Chem-Vac™ Chem-Cap® valves (A) instead of the CG-468-02 greased glass stopcocks on all of the 4-ports.
  • Four port vacuum manifold
  • Double bank
  • Left handed trap assembly
The end cap on the vacuum line is a #30 o-ring joint while the inert gas line has a #9 o-ring joint instead of the 14/20 standard taper joint. All gauging ports (B) are constructed using #9 o-ring joints and are supplied complete with adapters from the o-ring joint to 9 mm OØ tubulation for use with ³/₈" Cajon® fittings. Line uses the same main and pre-trap as the AF-0450 line.
Complete system consists of one 4-port vacuum/inert manifold, one pre-trap, one main trap assembly, one main trap body, two CG-150-05 #28 S.S. pinch clamps, one CG-150-08 #50 S.S. pinch clamp, one CG-150-21 #75 S.S. pinch clamp, four CG-150-04 #18-L S.S. pinch clamps, one AF-0451-05 #9 o-ring joint cap stopper, one AF-0451-21 #30 o-ring joint cap stopper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty