Would you like to log in again?
Mobile Phase HPLC Cap Systems, Chemglass
Supplier: Chemglass
CG-1167-12 CG-1167-11 CG-1167-13 CG-1167-14
76333-336EA 1121.51 USD InStock
76333-336 76333-332 76333-334 76333-330
Mobile Phase HPLC Cap Systems, Chemglass
Closures Safety Caps and Safety Waste Caps
The new and improved CG-1167 HPLC Cap system provides the safest and most economical way of storing high performance, low pressure liquid chromatography mobile phase, sparging preventing bubbles from entering the system as well as collecting sample waste.
The bottle is blue tinted plastic coated flat bottom, which provides additional safety by containing the glass fragments from accidents and allowing enough time to transfer samples or discarding safely.
Delivery information: Plastic Coated Glass Reservoir (1), 3-Hole Cap Assembly (1),
Combination Inlet/Sparger Filter 2 um (1), CG-1164-N-02 Flangless Nuts for 1.8" Tubing (2), CG-1164-F-02 Flangeless Ferrule for ¹/₈" Tubing (2), CG-1166-10 Tefzel Plug (1), CG-1164-02 ¹/₈" PTFE Tubing n 10' long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty