Would you like to log in again?
Support, Distillation Carts, Dual, 5 to 20 L, Chemglass
Supplier: Chemglass
CG-1968-D-102 CG-1968-D-100
76333-382EA 4522.6 USD InStock
76333-382 76333-384
Support, Distillation Carts, Dual, 5 to 20 L, Chemglass
Carts
Distillation receiver carts for use with our 10 L through 100 L reactor systems.
  • Glassware can be quickly installed and removed by one person
Heavy Duty support cart can be used for round bottom flasks or cylindrical distillation receivers. Rear of cart incorporates ½" stainless steel lattice for mounting support clamps or accessories.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty