Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Drain Valve, Zero Dead Space, GL Heavy Duty Threads, Pilot Plant, 2" Beaded Pipe, Chemglass
Supplier: Chemglass
CG-1968-GL-041 CG-1968-GL-040
76333-386EA 1565.5 USD InStock
76333-386 76333-388
Drain Valve, Zero Dead Space, GL Heavy Duty Threads, Pilot Plant, 2" Beaded Pipe, Chemglass
Valves Line Valves
The “Zero Dead Space” flush bottom valve is designed to prevent solids from collecting in the bottom of the vessel.
  • Glass body features a heavy duty GL style glass thread
  • Auto-stop when opening the valve
  • Anodized aluminum control knob is laser etched with directional arrows for opening and closing the valve
  • Zero dead space
  • Perfluoro O-rings on tip and plunger
The PTFE drain valve plunger is supplied with a Perfluoro and a TFE encapsulated o-ring for the tip and two Perfluoro o-rings installed on the shaft. The drain valve is connected to the reactor by a unique 2" beaded pipe valve seat that is sealed to the reactor bottom. Our valve permits total drainage of the vessel contents. The sidearm has a 1" beaded pipe fitting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty