Would you like to log in again?
Micro Graham Condenser, Chemglass
Supplier: Chemglass
CG-MR-50C2
76333-428EA 368.29 USD InStock
76333-428
Micro Graham Condenser, Chemglass
Condensers
Micro Graham condenser with 101 mm OAL, GL-14 top thread, 10/18 lower inner joint, and GL-14 thread inlet/outlet.
Delivery information: Supplied complete with caps with apertures and hose connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty