Would you like to log in again?
Double Port Adapters, M18, for Bioreactors, Chemglass
Supplier: Chemglass
CLS-1380-57
76333-448EA 427.04 USD InStock
76333-448
Double Port Adapters, M18, for Bioreactors, Chemglass
Fittings and Connectors
Adapter fits into the headplate and has two connections that are 4 mm OØ to fit precision tubing size 16. Inside the reactor, the 4 mm tubes are 1.25" long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty