Would you like to log in again?
Sampling Pipes, Harvest Tubes, Large Diameter, M18, for Bioreactors 2 - 5 L, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CLS-1380-70
76333-450EA 308.09 USD InStock
76333-450
Sampling Pipes, Harvest Tubes, Large Diameter, M18, for Bioreactors 2 - 5 L, Chemglass
Samplers Fluid Samplers
Pipe will fit a 2 to 5 liter reactor and has a lower diameter of ¹/₂" and a length of 212 mm into the reactor. The serrated hose connection is also ¹/₂" (12.7 mm) OØ. Can also be used as a quick way to harvest material due to its size.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty