Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dura-Mag™ Stirrers, Magnetic, 9-Position, 230 V

Supplier: Chemglass
CLS-4100-09E CLS-4100-A9E
76333-486EA 5984.9 USD InStock
76333-486 76333-476
Dura-Mag™ Stirrers, Magnetic, 9-Position, 230 V
Stirrers Magnetic Stirrers, Multi-position
The Chemglass life sciences nine-position magnetic stirrer is a rugged design for ²⁴/₇ laboratory or production applications.

  • 230 V model
  • Trouble-free brushless motor
  • Integrated power supply, for 100 to 220 V service
  • Heavy duty magnets and drive system
  • Trouble-free non-slip toothed Kevlar belt design with easy replacement
  • Stainless steel stirrer plate cover
  • Speed range from 25 to 400RPM with clockwise control
  • Dimensions are 16.75W×19.25D×3.5"H with the front to back depth measurement including power cord and control knobs

The small footprint is ideal for CO² incubators with maximum capacity of (9) 500 ml spinner vessels. Optional factory installed motion alarm available.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (76333-484) is also available, please see separate product page.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty