Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnetic Hot Plate Stirrer, Analog, 135 mm Plate, Velp

Supplier: VELP Scientifica
CG-1996-V10
76333-670EA 1077.17 USD InStock
76333-670
Magnetic Hot Plate Stirrer, Analog, 135 mm Plate, Velp
Stirrers Magnetic Hotplate Stirrers
Chemglass 135 mm round top ANALOG hot plate stirrer system.

  • The illuminated heating switch on/off prevents unintentional heat up
  • The illuminated heat icon informs the operator when the top plate temperature is above 50 °C
  • Resistant die-cast housing (IP) for maximum protection of the internal part of the instrument
  • The fast heating CerAITop plate resist to chemicals and scratches ensuring long life span
  • The stirring speed ramps up gently avoiding spillage risks

Hotplates feature many exclusive features and improved safety enhancements. The 135 mm diameter heater top is scratchproof and chemically-resistant. The back corner of the housing has an M8 female thread to accept a vertical support rod. 

Certifications: Hot plates have been designed and manufactured in compliance with the following standards: CE, CSA and UL.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty