Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cloud-Enabled Magnetic Hotplate Stirrer with Timer, AREX-6 CONN.PRO AREX-6 with Probe, Velp

Supplier: VELP Scientifica
CG-1997-W-50
76333-684EA 1531.27 USD InStock
76333-684
Cloud-Enabled Magnetic Hotplate Stirrer with Timer, AREX-6 CONN.PRO AREX-6 with Probe, Velp
Stirrers Magnetic Hotplate Stirrers
Velp Wifi enabled 135 mm round top DIGITAL magnetic hotplate stirrer system with built in timer.

  • High contrast, bright white LED
  • Powerful magnet: industry leading magnetic strength
  • Heating performance: fast precise heating/regulation up to 370 °C
  • Heat control modes: fine and fast
  • CerAlTop™ aluminum alloy and ceramic coating, 135 mm diameter
  • Three-year warranty

Velp Wifi enabled 135 mm round top DIGITAL magnetic hotplate stirrer system with built in timer. Hotplates feature many exclusive features and improved safety enhancements. The 135 mm diameter heater top is scratch proof and chemically resistant. The integrated TIMER provides an additional layer of safety ensuring the hotplate heater is "off" after the elapsed time (settable from 1 min to 99 h: 59 min). TRU-RPM stirring provides very accurate mixing speeds through the entire RPM range from 30 to 1700 rpm. The back corner of the housing has an M8 female thread to accept a vertical support rod. The updated interface, with easy to read, illuminated icons, create a very user-friendly experience. New and improved knobs and buttons assure simple and intuitive control of the magnetic hotplate stirrer.

Certifications: Hot plates have been designed and manufactured in compliance with the following standards CE, CSA and UL.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty