Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Stirring Shafts, Polished, 10 mm, Three-Blade, Glass Agitator, Retreating Blade, Clockwise, Chemglass
Supplier: Chemglass
CG-2079-C-50 CG-2079-C-60 CG-2079-C-58 CG-2079-C-52 CG-2079-C-54 CG-2079-C-56
76333-710EA 220.35 USD InStock
76333-710 76333-712 76333-718 76333-708 76333-714 76333-716
Stirring Shafts, Polished, 10 mm, Three-Blade, Glass Agitator, Retreating Blade, Clockwise, Chemglass
Stirrers Stirring Shafts
Shaft is precision ground and polished to fit our CG-2077-H, CG-2077-G and CG-2077-U style 10 mm PTFE bearings.
  • Stirrer shafts are designed for clockwise rotation
Designed to be used with cylindrical type reactors. The curved glass blades are spaced 120° apart and angled upward. 
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty