Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Replacement Stirrer Shaft, 25.4 mm, for Reactor Size 30 L, Chemglass
Supplier: Chemglass
CG-2097-33
76333-722EA 787.47 USD InStock
76333-722
Replacement Stirrer Shaft, 25.4 mm, for Reactor Size 30 L, Chemglass
Stirrers Stirring Shafts
Precision ground and polished stirrer shaft for use with Chemglass process reactors.
Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a ³/₈" wide flat.
Caution: Note: cg-2097-75 and cg-2097-100 are made from heavy wall tubing and have indents for agitator positioning. Indents are spaced every 10" from the bottom agitator.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty