Would you like to log in again?
Agar Deep, 1.5% (Water Agar), Hardy Diagnostics
Supplier: Hardy Diagnostics
Q01
76334-636EA 1136.92 USD InStock
76334-636
Agar Deep, 1.5% (Water Agar), Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Hardy Diagnostics Agar, 1.5% (Water Agar) is used in the Kirby-Bauer alternative agar-overlay method of inoculation for antimicrobial disk diffusion susceptibility testing and for microbiological procedures that require an additive-free solid medium, such as in performing colony counts, testing antimicrobial disk potency, or in performing the modified fungal slide culture method.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty