Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GMA 200-MW4 Series Controllers, Fixed Systems Gas Detection, GfG Instrumentation

Supplier: GfG Instrumentation
2200-093 2200-092 2200-094
76334-850EA 2122.5 USD InStock
76334-850 76334-854 76334-852
GMA 200-MW4 Series Controllers, Fixed Systems Gas Detection, GfG Instrumentation
Gas Detection Systems
The GMA 200-MW4 Controller is a high-performance gas measurement and warning system. The self-contained, IP 65 wall-mounted housing facilitates cost-effective and space-saving installation.

  • IP 65 wall-mounting housing, with integral audible and LED alarms
  • Up to four transmitters per GMA 200-MW4 controller in any combination
  • Flexible, reliable and economic
  • Six relays, four are programmable
  • Clearly structured, easy-to-use push-button interface
  • Backlit graphic LCD changes color to indicate alarm
  • Connect to other measurement devices with 4 to 20 mA output
  • Wall mounted dimensions approximately 8.125×7×2.25"

GMA 200-MW4 has a clearly structured, easy-to-use push-button interface and includes a backlit graphic LCD that changes color to indicate alarm.

Certifications: Applied for in accordance with ATEX 94/9/EC.

Ordering information: Sensors sold separately; contact your Avantor representative for more information.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty