Would you like to log in again?
IR 29 i Series Fixed Transmitters, Intrinsically Safe, Fixed Systems Gas Detection, GfG Instrumentation
Supplier: GfG Instrumentation
2901-21-004 2901-23-004 2901-37-004 2901-09-004 2901-99-004 2901-50-004
76334-882EA 2978.89 USD InStock
76334-882 76335-066 76334-880 76334-876 76334-884 76334-878
IR 29 i Series Fixed Transmitters, Intrinsically Safe, Fixed Systems Gas Detection, GfG Instrumentation
Gas Detection Systems
The IR 29 i is the first intrinsically safe IR gas transmitter with patented GfG 4-beam 4-wavelength technology. Because the IR 29 i is intrinsically safe, there is no need to run transmitter cabling through expensive explosion proof conduit.
  • ATEX: II II 1G Ex ia IIC T4 Ga
  • Intrinsically safe IR i transmitter for combustible gas measurement
  • Certified for Ex Zone 0 hazardous locations
  • Patented infrared technology (NDIR)
  • Temperature, moisture and pressure compensation
  • Energized IR sensor head can be replaced in Ex Zone 0 areas
  • Maintenance and service-friendly
The IR 29 i is the first IR gas transmitter that can be installed and used in “Ex Zone 0” hazardous locations in which ignitable concentrations of combustible gas or vapor are assumed to be present at all times. Use of the SB1 safety barrier (sold separately) protects the ATEX Certified system from the possibility of the release of energy that could be the cause of ignition.
Unlike traditional LEL sensors, IR 29 transmitters do not need the presence of oxygen to detect gas. Since they are not affected by sensor poisons they don’t require periodic adjustment to make up for lost sensitivity. The IR 29 continuously monitors its functions for proper performance, and can even identify and communicate required service procedures.
Certifications: ATEX: II II 1G Ex ia IIC T4 Ga. Intrinsically safe IR i transmitter for combustible gas measurement. Certified for Ex Zone 0 hazardous locations.
Ordering information: SB1 safety barrier is sold separately. For other available accessories or additional information, contact your Avantor representative and see the attached datasheet and manual.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty