Wherever hazards are expected, the EC 28 transmitter and GfG’s proven monitoring systems are the right choice for reliable surveillance. The ATEX-certified design, an integrated, Ex-proof buzzer, and bright alarm LEDs provide the highest safety in hazardous areas by including visual and audible alarms (optional). The gas alarm not only alerts the person in the control room of hazards, but also the person on-site.
- Increased safety design to reduce installation cost in potentially hazardous locations
- Non-intrusive AutoCal® calibration adjustment
- Uses electrochemical detection principle
- Output signal: 4 to 20 mA
- Available with MODBUS
Available in seven configurations:
Blind (no display)
Display
Display with local alarms
Display with local alarms and relays
Blind (no display) with MODBUS
Display with MODBUS
Display with local alarms and MODBUS
Smart sensor technology allows a user to install the transmitter or to replace a sensor within a few seconds – just plug the sensor into the transmitter. Adjustments are done directly at the transmitter using touch keys or with the RC2 remote control (one-person calibration).
The remote control display is always identical to that of the transmitter, making inspection, service, and calibration considerably simple. In addition, one remote control can be used for several transmitters, which helps cut costs and increase efficiency.
Certifications: ATEX-labelling: II 2G EEx em [ib] IIC T4 CE 0158.
EC-Type Examination Certificate: BVS 04 ATEX E 132 x.
Features: no safety barrier necessary.
Accessories information: MODBUS communication interface available for all transmitter configurations except Display/Alarm/Relay (004). Add “M” to the part number (e.g. 2811-17-001M) and add an additional fee, contact your representative in this case.
Ordering information: For all applications above –4 degrees farenheit, GfG recommends the standard NMH₃ sensor configuration. Please contact the manufacturer for proper selection of NH₃ sensors. For available accessories or additional information, contact your Avantor representative and see the attached datasheet and manual.