Would you like to log in again?
KLEENGUARD® A20 Breathable Particle Protection Lab Coats with Serged Seams, Kimberly-Clark Professional
Supplier: Kimberly-Clark
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KleenGuard®
35622 35624 35619 35621 35623 35625 35618 35620
76335-134CS 336.12 USD InStock
76335-134 76335-136 76334-620 76334-626 76334-616 76334-614 76334-628 76334-618
KLEENGUARD® A20 Breathable Particle Protection Lab Coats with Serged Seams, Kimberly-Clark Professional
Workwear
KLEENGUARD® Personal Protective Equipment (PPE) enables rather than inhibits, allowing for maximum productivity and safety for both employees and facilities.
  • Designed to help protect against dirt, grime and dust
  • Made with breathable Microforce barrier fabric
  • Garments feature serged seams, four hook & loop closures, and elastic wrists
  • Knee-length lab coats do not have pockets
  • Passes NFPA 99 for anti-static materials
These lab coats are designed to move with the wearer, while helping protect against hazardous and non-hazardous dust. These protective garments offer superior particle holdout, coupled with unmatched comfort attributes. The A20 fabric is comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles.
This knee-length model comes in a variety of sizes and features serged seams, four hook and loop closures, and elastic wrists. A popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications or as decontamination modesty garments.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty