Would you like to log in again?
ProCulture® Complete Spinner Flask with Angled Sidearms and Magnetically Driven Vortex Impeller, SP Industries
Supplier: SP Industries
ProCulture®
BP-3400-104 BP-3400-106 BP-3400-116 BP-3400-100 BP-3400-112 BP-3400-102 BP-3400-108 BP-3400-114 BP-3400-110
76335-220EA 1196.8 USD InStock
76335-220 76335-222 76335-212 76335-224 76335-214 76335-226 76335-216 76335-228 76335-218
ProCulture® Complete Spinner Flask with Angled Sidearms and Magnetically Driven Vortex Impeller, SP Industries
Flasks Spinner Flasks
Designed to save time and increase cell production, ProCulture® spinner flasks have internal baffles for enhanced agitation and a magnetically driven vortex impeller that provides gentle top-to-bottom stirring.
  • Sidearm openings large enough for 50 ml pipettes
  • Flask base has central bump to prevent material from accumulating at the bottom center
  • Entire assembly made of FDA approved materials
  • Cap made from Polybutylene terephthalate (PBT), red rubber, Ultem®
  • Flask is type 1, class A borosilicate glass construction
  • All components are autoclavable
The impeller eliminates dead spots and extends above the media surface to increase aeration. It is made from PTFE, silicone, polysulfone, borosilicate glass.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty