Would you like to log in again?
Sealing Tubes, Beckman Coulter
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
355630 355630 355630
BK355630PK 320.61 USD InStock
BK355630 CABK355630 76335-408
Sealing Tubes, Beckman Coulter
Tubes Centrifuge Tubes
These sealing tubes are designed for use in specific rotor types and applications.
  • Heat-sealed tubes used in swinging bucket, vertical tube, near vertical tube, and most fixed-angle rotors
  • Bell-top style simplifies removal of materials that float during centrifugation
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty