Would you like to log in again?
TUBE THICKWALL ROUND BTM PP 38.5 ML BX25
Supplier: BECKMAN COULTER
Tube Thickwall Round Btm PP 38.5 Ml BX25^ Open Top Polypropylene Round Bottom tube, Thickwall, Platform: Floor Prep, Optical Property: Transparent, For use in rotors: Type 70 Ti, Type 50.2 Ti, SW 32 Ti, SW 28, Nominal Size: 25 x 89mm, Nominal Capacity: 38.5mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE THICKWALL ROUND BTM PP 38.5 ML BX25 76335-432 355642 Box of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty