Would you like to log in again?
OptiSeal Ultracentrifugation Tubes, Beckman Coulter
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
362183 361621 362185 362183 361623 361627 362185 361623 362181 361621 362181
76335-440BX 356.98 USD InStock
76335-440 76335-420 CA76333-076 76335-642 76335-412 CA76333-094 76335-438 76335-648 CA76333-744 CA76333-070 CA76333-092
OptiSeal Ultracentrifugation Tubes, Beckman Coulter
Tubes Centrifuge Tubes
The OptiSeal ultracentrifugation tubes provide a fast, reliable seal every time. Seal with a finger-touch and without tools, heat, or closure verification. Wide necks offer effortless filling and fraction collection.
  • Easy-to-Use
  • Sliceable/Puncturable
  • Wide selection
Whether you are helping to solve complex biological problems, investigating the causes of diseases or finding potential new cures, you need consumables that meet your workflow needs.
For virtually effortless sealing, OptiSeal tubes offer an excellent option. You simply insert the tube plug and press, and an O-ring seals securely against the tube's inner surface. During centrifugation, the combination of g-force and hydrostatic pressure ensures an effective seal. Engineered for reliability, there are no tools or mechanical parts to maintain. Finger pressure is all that is needed.
Caution: Please always check the centrifuge rotor manual for the list of compatible tubes and adapters.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty