Would you like to log in again?
TUBE THICKWALL ROUND BTM PP 3.5 ML BX25
Supplier: BECKMAN COULTER
Tube Thickwall Round Btm PP 3.5 Ml BX25^ Open Top Polypropylene Round Bottom tube, Thickwall, Platform: Tabletop UCF, Optical Property: Transparent, For use in SW 55 Ti rotor, Nominal Size: 13 x 51mm, Nominal Capacity: 3.5mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE THICKWALL ROUND BTM PP 3.5 ML BX25 76335-442 349623 Box of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty