Would you like to log in again?
TUBE QUICK-SEAL POLYPROPYLENE 4 ML PK50
Supplier: BECKMAN COULTER
Tube Quick-Seal Polypropylene 4 Ml PK50^ Quick-Seal* Polypropylene Tube, For Use In: Ultracentrifuge Rotors: TLN-100, Platform: Tabletop UCF, Round-Top, Nominal Capacity: 4 mL, Nominal Size: 13 x 38mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE QUICK-SEAL POLYPROPYLENE 4 ML PK50 76335-496 358980 Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty