Would you like to log in again?
TUBE QUICK-SEAL ROUND BOTTOM 27ML BX50
Supplier: BECKMAN COULTER
Tube Quick-Seal Round Bottom 27Ml BX50^ Quick-seal* Ultra-clear Round Bottom tube, bell top, Platform: Floor Prep, Optical Property: Transparent, For use in rotors: Type 70 Ti, Type 50.2 Ti, VTi 50, Nominal Size: 25 x 64mm, Nominal Capacity: 27mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE QUICK-SEAL ROUND BOTTOM 27ML BX50 76335-502 344323 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty