Would you like to log in again?
KIT G-MAX QCKSEAL POLYPRPYLNE 3.5ML BX50
Supplier: BECKMAN COULTER
Kit G-Max Qckseal Polyprpylne 3.5Ml BX50^ Quick-Seal* Polypropylene Round Bottom tube, g-Max Kit, Platform: Floor Prep, for use in TYPE 100 Ti rotor. Nominal Capacity: 3.5ml, Nominal Size: 13 x 32 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT G-MAX QCKSEAL POLYPRPYLNE 3.5ML BX50 76335-508 363961 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty