Would you like to log in again?
VWR® LTS Compatible Adjustable Volume Pipettors
Supplier: VWR
VWR®
30537533 30537531 30551490 30537529 30551494 30551488 30551493 30537530 30551491 30537532 30551492 30551489 30537535 30537528 30537534
76382-698EA 1030.67 USD InStock
76382-698 76335-572 76382-696 76382-694 76335-742 76382-706 76382-704 76382-702 76382-700 76335-570 76382-708 76335-568 76335-744 76335-746 76335-748
VWR® LTS Compatible Adjustable Volume Pipettors
Pipettes Micropipettes
Patented tip ejection technology reduces fatigue and makes the VWR® LTS compatible pipettor a top choice to improve your lab’s ergonomics.
  • Calibrated to ISO 8655 standards with Biotix xTIP4 tips
  • Fully compatible with Rainin LTS tips
  • Light spring forces produce a smooth and controlled aspiration with perfect balance
  • Padded ejector button to reduce hand vibrations during tip ejection
  • Simple volume adjustment with a secure lock to prevent volume drift
  • Available in single, 8, and 12-channel models with a symmetrical design for ambidextrous use
The VWR® LTS compatible pipettor uses durable and lightweight components that deliver a precise and accurate liquid transfer while maintaining optimal hand comfort.
Certifications: ISO 8655 certified. Calibration certificate included in the box with each pipette. Unique serial ID number under the finger hook for GLP/GMP compliance.
Caution: Rainin and LTS are a trademark of Mettler-Toledo, Rainin LLC.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty