Would you like to log in again?
KIT TUBE QUICKSEAL POLYPROPLN 6.3ML BX50
Supplier: BECKMAN COULTER
Kit Tube Qckseal Polyprpylne F/6.3MlBX50^ Quick-Seal* Polypropylene Round Bottom tube kit, Platform: Floor Prep, for use in rotors: Type 90 Ti, Type 70.1 Ti. Nominal Capacity: 6.3ml, Nominal Size: 16 x 45 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT TUBE QUICKSEAL POLYPROPLN 6.3ML BX50 76335-578 357334 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty