Would you like to log in again?
KIT TUBE QUICKSEAL ROTOR 1 X 3-1/2
Supplier: BECKMAN COULTER
Kit Tube Quickseal Rotor 1 X 3-1/2^ Quickseal* Tube Kit, Includes 2 boxes 342414 Polypropylene Tubes, 2 boxes 344326 Ultra-Clear Tubes, 12 Spacers 342699, 1 each 348124 Tube Rack, 306812 Spinkote Lubricant, and 335148 Vacuum Grease, Cordless tube topper kit required.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
KIT TUBE QUICKSEAL ROTOR 1 X 3-1/2 76335-636 348178 Kit of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty