Would you like to log in again?
TUBE ULTRA-CLEAR ROUND BOTTOM 0.8ML BX50
Supplier: BECKMAN COULTER
Tube Ultra-Clear Round Bottom 0.8Ml BX50^ Open Top Ultra-clear Round Bottom tube, Thinwall, Platform: Floor Prep, Optical Property: Transparent, For use in SW 55 Ti rotor, Nominal Size: 5 x 41mm, Nominal Capacity: 0.8mL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE ULTRA-CLEAR ROUND BOTTOM 0.8ML BX50 76335-646 344090 Box of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty