Would you like to log in again?
TUBE POLYETHYLENE THICKWALL 175 UL BX100
Supplier: BECKMAN COULTER
Tube Polyethylene Thickwall 175 Ul BX100^ Polyethylene Tube, Platform: Airfuge, Closure: Open-Top Thickwall, For Use In Ultracentrifuge Rotors TLS-55 (Part Numbers 346936, 346134) A-110 (Part Number 347596) A-100/30, Nominal Capacity: 175 uL, Nominal Size:5 x 20mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBE POLYETHYLENE THICKWALL 175 UL BX100 76335-658 343622 Box of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
DOCUMENTATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty