Would you like to log in again?
Certified Free and Sterile Ultracentrifugation Tubes, Beckman Coulter
Supplier: Beckman Coulter
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
C14301 C14279 C14307 C14286 C14304 C14285 C14283 C14277 C14287 C13926 C14299 C14281 C14302 C14277 C18669 C14282 C14278 C14281 C14293 C14297 C14306 C14302 C14288 C14296 C14278 C14286 C14296 C14304 C14305 C14294 C14295 C14297 C14290 C14307 C14294 C14298 C14290 C14298 C14288 C14292 C14283 C14282 C18669 C14280 C14291 C14303 C14303 C14287 C14279 C14289 C14299 C14293 C14285 C14295 C14289 C18674 C14305 C13926 C14306 C14280 C18674 C14292
76335-730BX 671.05 USD InStock
CA76333-138 CA76333-176 76335-730 CA76333-298 76335-610 CA76333-178 76335-732 CA76333-172 CA76333-174 CA76333-170 76335-722 76335-724 76335-726 76335-606 76335-728 76335-608 CA76333-168 CA76333-164 76335-620 CA76333-166 76335-622 CA76333-160 CA76333-162 76335-612 76335-614 76335-616 76335-618 CA76333-158 CA76333-314 76335-630 CA76333-154 76335-632 76335-710 CA76333-156 CA76333-310 CA76333-150 CA76333-152 76335-708 CA76333-506 CA76333-308 76335-700 76335-624 76335-702 76335-626 CA76333-304 76335-704 76335-628 CA76333-504 76335-706 CA76333-306 CA76333-146 CA76333-300 CA76333-302 CA76333-148 CA76333-142 CA76333-144 76335-720 CA76333-140 76335-634 76335-712 76335-714 76335-718
Certified Free and Sterile Ultracentrifugation Tubes, Beckman Coulter
Tubes Centrifuge Tubes
These tubes help protect your sample from degradation and contamination while solving complex biological problems, investigating the causes of diseases or finding potential new cures. They provide sample protection when it matters most.
  • Maximize consistency
  • Prevent yield loss
  • Ready-to-go, right out of the bag
Certified free: These tubes exhibit no detectable DNA, DNase, RNase, PCR inhibitors, or endotoxins. (Based on sample results below detectable limits.)
• Ideal for genomics, exosomes, and proteomics workflows
• Prevents sample degradation and contamination
• Double-bagged to prevent contamination during clean room transfer
• Improve QC data quality
Certified free + Sterile: Beckman sterile ultracentrifuge tubes are sterilized via ethylene oxide.
• Ideal for protein prep and virus purification workflows
• Includes all advantages of being certified free
• Pre-sterilized ready-to-use for improved workflow efficiency
• Eliminate need for developing sterilization protocolol
• Packaged in workflow based quantities
• Sterilized and packaged compliant with ISO 11135 and 11607
Unique applications and workflows have varying requirements for the presence of viable and nonviable biologic materials. Samples may require consumables verified as certified free, sterile, or both. Choosing the right tube, provides your sample or experiment the best possible outcome.
Certified free: Genomics, DNA/RNA sequencing, exosomes and proteomics.
Certified free + Sterile: Microbiology, gene therapy, biotherapeutics, molecular biology, cell culture, plant/animal derived products and algae.
Caution: Please always check the centrifuge rotor manual for the list of compatible tubes and adapters.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty