Would you like to log in again?
Corepoint Scientific™ Controlled Room Temperature (CRT) Climatic Chambers, Horizon Scientific
Supplier: Horizon Scientific
CRTPR011WWW/0 CRTPR051WWG-0 CRTPR051WWWLH0 CRTPR011WWWLH/0 CRTPR101WWG-0 CRTPR031WWG/0 CRTPR011WWG/0 CRTPR061WWG-0 CRTPR051WWW-0 CRTPR101WWW-0 CRTPR031WWW/0 CRTPR051WWGLH0 CRTPR011WWGLH/0
76336-704EA 4941.48 USD InStock
76336-704 76336-702 76336-688 76336-666 76336-700 76336-690 76336-686 76336-698 76336-684 76336-696 76336-682 76336-694 76336-692
Corepoint Scientific™ Controlled Room Temperature (CRT) Climatic Chambers, Horizon Scientific
Testing Chambers Climatic Chambers
These CRT cabinets provide controlled room temperature environments for storing sensitive substances that require precise temperature management.
  • Integrated temperature alarms
  • Keyed door locks
  • Forced draft air circulation
  • Probe access
  • Precise microprocessor temperature control
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These specialized cabinets are ideal for scientific and medical applications. Both built-in and freestanding configurations are available. Built-in units are designed with front-facing ventilation systems, allowing easy installation into a bench or countertop. Freestanding units require a 4-inch margin on all sides for adequate ventilation. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76336-666, 76336-682, 76336-684, 76336-686, 76336-688, 76336-690, 76336-692, 76336-694, 76336-696, 76336-698, 76336-700, 76336-702, and 76336-704
Ordering information: All units have a two-year parts and labor warranty. Selected models come with an additional three-year compressor parts warranty. (Please refer to unit datasheets to confirm).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty