Would you like to log in again?
CRUCIBLE 5 % GOLD 95 %PLATINUM STD 150ML
Supplier: XRF Scientific
Crucible 5 % Gold 95 %Platinum Std 150Ml^ Crucible, Reinforcing of the rim gives the Crucible a more robust character enabling regular handling and reducing rim deformation, To optimize the labware for the chosen processes, are made out of 5% gold 95% platinum standard shape, Size: 150 ml capacity
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRUCIBLE 5 % GOLD 95 %PLATINUM STD 150ML 76336-734 5AUPTCR150L Each (150ml) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty