Would you like to log in again?
CRUCIBLE 5%GOLD-95% PT WIDE SHAPE 40ML
Supplier: XRF Scientific
Crucible 5%Gold-95% Pt Wide Shape 40Ml^ Crucible, To optimize the labware for the chosen processes in respect to temperature, stability, susceptibility to oxidation, easy cleaning and lifetime performance we have several materials, are made out of 5% gold 95% platinum wide shape, Size: 40 ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CRUCIBLE 5%GOLD-95% PT WIDE SHAPE 40ML 76336-778 5AUPTLWC40L Each (40ml) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty